ENA308583294 Enamine

sodium 2-(2,3-dihydro-1H-indol-1-yl)-2-phenylacetate

  • MDL number MFCD17167210
  • Cas Number : 1258639-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68818

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68818
Availability details for
Quantity
  • Availability
  • Add to Cart