ENA308583298 Enamine

N'-(2-carbamoylethyl)butanediamide

  • MDL number MFCD17167215
  • Cas Number : 1258650-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68858
Availability details for
Quantity
  • Availability
  • Add to Cart