ENA308583299 Enamine

2-(piperazin-1-yl)-1-(piperidin-1-yl)propan-1-one dihydrochloride

  • MDL number MFCD17167217
  • Cas Number : 1258639-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68867
Availability details for
Quantity
  • Availability
  • Add to Cart