ENA308583301 Enamine

1-N,3-N-dicyclopropyl-2-nitrobenzene-1,3-diamine

  • MDL number MFCD17167220
  • Cas Number : 1258650-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68895
Availability details for
Quantity
  • Availability
  • Add to Cart