ENA308583302 Enamine

N-methyl-3-(trifluoromethoxy)aniline hydrochloride

  • MDL number MFCD17167222
  • Cas Number : 1201824-14-7
  • Molecular Weight : 227.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68908

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-68908
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart