ENA308583303 Enamine

3-[(5-methyl-1,2,4-oxadiazol-3-yl)methyl]piperidine hydrochloride

  • MDL number MFCD17167223
  • Cas Number : 1258650-44-0
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68913
Availability details for
Quantity
  • Availability
  • Add to Cart