ENA308583312 Enamine

2-(4-chlorophenyl)-2-(2,2-dimethylmorpholin-4-yl)acetonitrile

  • MDL number MFCD17167236
  • Cas Number : 1258650-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68990
Availability details for
Quantity
  • Availability
  • Add to Cart