ENA308583315 Enamine

2-[(3-aminophenyl)(benzyl)amino]acetamide

  • MDL number MFCD17167239
  • Cas Number : 1258650-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69036
Availability details for
Quantity
  • Availability
  • Add to Cart