ENA308583328 Enamine

2-chloro-N-[(3,4-dimethoxyphenyl)methyl]-N-methylpropanamide

  • MDL number MFCD11547004
  • Cas Number : 1094533-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28160
Availability details for
Quantity
  • Availability
  • Add to Cart