ENA308583329 Enamine

N-[(4-bromophenyl)methyl]-2-chloro-N-methylpropanamide

  • MDL number MFCD11643915
  • Cas Number : 1098347-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28170
Availability details for
Quantity
  • Availability
  • Add to Cart