ENA308583336 Enamine

1-(4-aminophenyl)-3-(propan-2-yl)urea

  • MDL number MFCD11167417
  • Cas Number : 70826-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29215
Availability details for
Quantity
  • Availability
  • Add to Cart