ENA308583344 Enamine

[(5-bromo-2-fluorophenyl)methyl](propan-2-yl)amine

  • MDL number MFCD09810839
  • Cas Number : 1016741-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-32156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32156
Availability details for
Quantity
  • Availability
  • Add to Cart