ENA308583347 Enamine

benzyl[(4-methylphenyl)methyl]amine

  • MDL number MFCD03210669
  • Cas Number : 55096-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32616
Availability details for
Quantity
  • Availability
  • Add to Cart