ENA308583348 Enamine

benzyl[1-(thiophen-2-yl)ethyl]amine

  • MDL number MFCD11140751
  • Cas Number : 1019534-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-32626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32626
Availability details for
Quantity
  • Availability
  • Add to Cart