ENA308583367 Enamine

(2,3-dihydro-1H-inden-5-yl)methanamine hydrochloride

  • MDL number MFCD11813965
  • Cas Number : 721968-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41205
Availability details for
Quantity
  • Availability
  • Add to Cart