ENA308583374 Enamine

1-[(2-methoxyphenyl)methyl]-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00114057
  • Cas Number : 133137-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44379
Availability details for
Quantity
  • Availability
  • Add to Cart