ENA308583377 Enamine

4-methyl-N1-(propan-2-yl)benzene-1,2-diamine

  • MDL number MFCD11645461
  • Cas Number : 1097810-17-7
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45053
Availability details for
Quantity
  • Availability
  • Add to Cart