ENA308583382 Enamine

4-fluoro-1-N-(propan-2-yl)benzene-1,2-diamine

  • MDL number MFCD06408848
  • Cas Number : 153877-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45059
Availability details for
Quantity
  • Availability
  • Add to Cart