ENA308583385 Enamine

N1-cyclopropyl-3-fluorobenzene-1,2-diamine

  • MDL number MFCD12149485
  • Cas Number : 1179208-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45077
Availability details for
Quantity
  • Availability
  • Add to Cart