ENA308583386 Enamine

2-chloro-N-[3-(cyclohexyloxy)propyl]acetamide

  • MDL number MFCD14588092
  • Cas Number : 91354-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46459
Availability details for
Quantity
  • Availability
  • Add to Cart