ENA308583388 Enamine

2-chloro-N-[2-(3-fluorophenoxy)ethyl]acetamide

  • MDL number MFCD16316213
  • Cas Number : 1226121-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-46756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46756
Availability details for
Quantity
  • Availability
  • Add to Cart