ENA308583403 Enamine

2-chloro-N-{2-[4-(propan-2-yl)phenoxy]ethyl}acetamide

  • MDL number MFCD16330117
  • Cas Number : 1225489-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-47662
Availability details for
Quantity
  • Availability
  • Add to Cart