ENA308583406 Enamine

2-chloro-1-[2-(thiophen-3-yl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD12798916
  • Cas Number : 1179119-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-47843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47843
Availability details for
Quantity
  • Availability
  • Add to Cart