ENA308583415 Enamine

N-(1-benzyl-1H-pyrazol-3-yl)-2-chloroacetamide

  • MDL number MFCD17167097
  • Cas Number : 1258641-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48271
Availability details for
Quantity
  • Availability
  • Add to Cart