ENA308583420 Enamine

N-[2-(2H-1,3-benzodioxol-5-yloxy)ethyl]-2-chloroacetamide

  • MDL number MFCD16331254
  • Cas Number : 1225816-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48514
Availability details for
Quantity
  • Availability
  • Add to Cart