ENA308583423 Enamine

N-[1-(2-chloroacetyl)piperidin-3-yl]acetamide

  • MDL number MFCD12762700
  • Cas Number : 1182969-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48886
Availability details for
Quantity
  • Availability
  • Add to Cart