ENA308583449 Enamine

4-chloro-6-ethylpyrimidine

  • MDL number MFCD09746254
  • Cas Number : 141602-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53289
Availability details for
Quantity
  • Availability
  • Add to Cart