ENA308583508 Enamine

tert-butyl N-(3-hydroxypropyl)-N-(propan-2-yl)carbamate

  • MDL number MFCD12781066
  • Cas Number : 873437-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61982
Availability details for
Quantity
  • Availability
  • Add to Cart