ENA308583512 Enamine

2-{[(tert-butoxy)carbonyl]amino}pentanoic acid

  • MDL number MFCD00270725
  • Cas Number : 521286-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62478

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62478
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart