ENA308583532 Enamine

tert-butyl 2-cyanopyrrolidine-1-carboxylate

  • MDL number MFCD01861225
  • Cas Number : 144688-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63964

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63964
Availability details for
Quantity
  • Availability
  • Add to Cart