ENA308583540 Enamine

(1S)-1-(5-chloro-2,4-dimethoxyphenyl)ethan-1-ol

  • MDL number MFCD09863715
  • Cas Number : 1175985-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64639
Availability details for
Quantity
  • Availability
  • Add to Cart