ENA308583541 Enamine

N-(cyclopropylmethyl)-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-5-amine

  • MDL number MFCD12039902
  • Cas Number : 1157795-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64788

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64788
Availability details for
Quantity
  • Availability
  • Add to Cart