ENA308583542 Enamine

(1R)-1-[2-(benzyloxy)phenyl]ethan-1-ol

  • MDL number MFCD09863700
  • Cas Number : 181962-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64908
Availability details for
Quantity
  • Availability
  • Add to Cart