ENA308583556 Enamine

2-({4H,5H,6H-cyclopenta[b]thiophen-2-yl}formamido)acetic acid

  • MDL number MFCD09813933
  • Cas Number : 1016533-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65458
Availability details for
Quantity
  • Availability
  • Add to Cart