ENA308583557 Enamine

1-[(3-aminophenyl)methyl]piperidine-3-carboxamide

  • MDL number MFCD09809240
  • Cas Number : 1016509-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65551
Availability details for
Quantity
  • Availability
  • Add to Cart