ENA308583561 Enamine

4-(hydroxymethyl)-2-iodo-6-methoxyphenol

  • MDL number MFCD00238615
  • Cas Number : 37987-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65776
Availability details for
Quantity
  • Availability
  • Add to Cart