ENA308583565 Enamine

1-(4-nitrophenyl)-1H-pyrazol-5-amine

  • MDL number MFCD11641431
  • Cas Number : 16459-44-2
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66077
Availability details for
Quantity
  • Availability
  • Add to Cart