ENA308583568 Enamine

phenyl(pyridin-3-yl)methanol

  • MDL number MFCD02942350
  • Cas Number : 6270-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66208

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-66208
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart