ENA308583574 Enamine

4-(4-nitrophenyl)-1H-pyrazole

  • MDL number MFCD04037972
  • Cas Number : 114474-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66376
Availability details for
Quantity
  • Availability
  • Add to Cart