ENA308583598 Enamine

N1-(pyridazin-3-yl)ethane-1,2-diamine

  • MDL number MFCD16767528
  • Cas Number : 193473-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67019
Availability details for
Quantity
  • Availability
  • Add to Cart