ENA308583608 Enamine

1-(2-methanesulfonylethyl)piperazine dihydrochloride

  • MDL number MFCD02684112
  • Cas Number : 870007-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67166
Availability details for
Quantity
  • Availability
  • Add to Cart