ENA308583624 Enamine

3-[(4-chlorophenyl)carbonyl]-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD03015442
  • Cas Number : 1042437-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67525
Availability details for
Quantity
  • Availability
  • Add to Cart