ENA308583629 Enamine

(1S)-1-(2-ethoxyphenyl)ethan-1-ol

  • MDL number MFCD09863755
  • Cas Number : 1212111-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67576
Availability details for
Quantity
  • Availability
  • Add to Cart