ENA308583631 Enamine

2-[(1,3-dimethyl-2,6-dioxo-2,3,6,9-tetrahydro-1H-purin-8-yl)sulfanyl]acetic acid

  • MDL number MFCD17480357
  • Molecular Weight : 270.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67596
Availability details for
Quantity
  • Availability
  • Add to Cart