ENA308583634 Enamine

3-(4-amino-2-fluorophenoxymethyl)benzonitrile

  • MDL number MFCD11526352
  • Cas Number : 1094859-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67643

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67643
Availability details for
Quantity
  • Availability
  • Add to Cart