ENA308583635 Enamine

4-(pyridine-4-amido)benzene-1-sulfonyl chloride

  • MDL number MFCD09815240
  • Cas Number : 89565-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-67646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67646
Availability details for
Quantity
  • Availability
  • Add to Cart