ENA308583637 Enamine

3-(3-fluorophenoxy)propanamide

  • MDL number MFCD11644015
  • Cas Number : 1094734-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67663
Availability details for
Quantity
  • Availability
  • Add to Cart