ENA308583638 Enamine

3-(2-fluorophenoxy)propanamide

  • MDL number MFCD11643184
  • Cas Number : 1097829-72-5
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67664
Availability details for
Quantity
  • Availability
  • Add to Cart