ENA308583640 Enamine

2-[(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)amino]butanoic acid

  • MDL number MFCD12071845
  • Cas Number : 1218314-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67792
Availability details for
Quantity
  • Availability
  • Add to Cart