ENA308583641 Enamine

4-chloro-2-(difluoromethyl)quinazoline

  • MDL number MFCD11193508
  • Cas Number : 1039312-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67828

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67828
Availability details for
Quantity
  • Availability
  • Add to Cart